missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EPGN Full-length ORF (AAI46548.1, 1 a.a. - 133 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00255324-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MALGVPISVYLLFNAMTALTEEAAVTVTPPITAQQADNIEGPIALKFSHLCLEDHNSYCINGACAFHHELEKAICRCFTGYTGERCEHLTLTSYAVDSYEKYIAIGIGVGLLLSGFLVIFYCYIRKRYEKDKISpecifications
AAI46548.1 | |
Liquid | |
255324 | |
EPGN (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MALGVPISVYLLFNAMTALTEEAAVTVTPPITAQQADNIEGPIALKFSHLCLEDHNSYCINGACAFHHELEKAICRCFTGYTGERCEHLTLTSYAVDSYEKYIAIGIGVGLLLSGFLVIFYCYIRKRYEKDKI | |
RUO | |
EPGN | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
41.58kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALGV3072/EPG/FLJ75542/PRO9904/epigen | |
EPGN | |
Yes | |
wheat germ expression system |