missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Gibco™ Human ENA-78 (CXCL5) (5-78 aa) Recombinant Protein, PeproTech®
Klik for at se tilgængelige muligheder
Mængde:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
Beskrivelse
Recombinant Human ENA-78 is an 8.0 kDa protein consisting of 74 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
The protein encoded by this gene is an inflammatory chemokine that belongs to the CXC chemokine family. This chemokine is produced concomitantly with interleukin-8 (IL8) in response to stimulation with either interleukin-1 (IL1) or tumor necrosis factor-alpha (TNFA). This chemokine is a potent chemotaxin involved in neutrophil activation.
Tekniske data
Tekniske data
| Adgangsnummer | P42830 |
| Til brug med (applikation) | ELISA, Functional Assay, Western Blot |
| Formulering | protein with no preservative |
| Gen-id (Entrez) | 6374 |
| Molekylvægt (g/mol) | 8 kDa |
| Navn | Human ENA-78 (CXCL5) (5-78 aa) |
| Mængde | 50 μg |
| Kilde | E. coli |
| Regulatorisk status | RUO |
| Endotoksin koncentration | <1 EU/ μg |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion