missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EFHD1 Partial ORF (NP_079478.1, 168 a.a. - 238 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_079478.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 80303 |
Molecular Weight (g/mol) | 33.44kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16144913
|
Abnova™
H00080303-Q01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16134913
|
Abnova™
H00080303-Q01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
EFHD1 is an EF-hand domain-containing protein that displays increased expression during neuronal differentiation (Tominaga and Tomooka, 2002 [PubMed 12270117]).[supplied by OMIM]
Sequence: GLMALAKLSEIDVALEGVKGAKNFFEAKVQALSSASKFEAELKAEQDERKREEEERRLRQAAFQKLKANFNSpecifications
NP_079478.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.44kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp781H0842/FLJ13612/MST133/MSTP133/PP3051 | |
EFHD1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
80303 | |
EFHD1 (Human) Recombinant Protein (Q01) | |
GLMALAKLSEIDVALEGVKGAKNFFEAKVQALSSASKFEAELKAEQDERKREEEERRLRQAAFQKLKANFN | |
RUO | |
EFHD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |