missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DUSP21 Full-length ORF (NP_071359.2, 1 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_071359.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 63904 |
Molecular Weight (g/mol) | 47.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16133293
|
Abnova™
H00063904-P01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16123293
|
Abnova™
H00063904-P01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the dual specificity phosphatase family, specifically the low molecular weight dual specificity phosphatase family. The encoded protein localizes to both the cytoplasm and the nucleus and functions to remove phosphate groups from phosphotyrosine and phosphothreonine residues
Sequence: MTASASSFSSSQGVQQPSIYSFSQITRSLFLSNGVAANDKLLLSSNRITAIVNASVEVVNVFFEGIQYIKVPVTDARDSRLYDFFDPIADLIHTIDMRQGRTLLHCMAGVSRSASLCLAYLMKYHSMSLLDAHTWTKSRRPIIRPNNGFWEQLINYEFKLFNNNTVRMINSPVGNIPDIYEKDLRTMISMSpecifications
NP_071359.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
47.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MTASASSFSSSQGVQQPSIYSFSQITRSLFLSNGVAANDKLLLSSNRITAIVNASVEVVNVFFEGIQYIKVPVTDARDSRLYDFFDPIADLIHTIDMRQGRTLLHCMAGVSRSASLCLAYLMKYHSMSLLDAHTWTKSRRPIIRPNNGFWEQLINYEFKLFNNNTVRMINSPVGNIPDIYEKDLRTMISM | |
RUO | |
DUSP21 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
63904 | |
DUSP21 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LMWDSP21/MGC149878 | |
DUSP21 | |
Recombinant | |
wheat germ expression system |