missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DUSP18 Full-length ORF (NP_689724.3, 1 a.a. - 188 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00150290-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138603

  • 4040.00 DKK / 25µg
Estimated Shipment: 03-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

DUSP18 is a member of the dual-specificity phosphatase (DSP) family (see DUSP1; MIM 600714), which catalyzes dephosphorylation of phosphotyrosine and phosphothreonine residues (Hood et al., 2002 [PubMed 12408986]).[supplied by OMIM]

Sequence: MTAPSCAFPVQFRQPSVSGLSQITKSLYISNGVAANNKLMLSSNQITMVINVSVEVVNTLYEDIQYMQVPVADSPNSRLCDFFDPIADHIHSVEMKQGRTLLHCAAGVSRSAALCLAYLMKYHAMSLLDAHTWTKSCRPIIRPNSGFWEQLIHYEFQLFGKNTVHMVSSPVGMIPDIYEKEVRLMIPL
Specifications

Specifications

NP_689724.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
47.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DSP18/DUSP20/LMWDSP20/MGC32658/bK963H5.1
DUSP18
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
150290
DUSP18 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTAPSCAFPVQFRQPSVSGLSQITKSLYISNGVAANNKLMLSSNQITMVINVSVEVVNTLYEDIQYMQVPVADSPNSRLCDFFDPIADHIHSVEMKQGRTLLHCAAGVSRSAALCLAYLMKYHAMSLLDAHTWTKSCRPIIRPNSGFWEQLIHYEFQLFGKNTVHMVSSPVGMIPDIYEKEVRLMIPL
RUO
DUSP18
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers