missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DSN1 Full-length ORF (AAH58899.1, 1 a.a. - 356 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079980-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16144693

  • 4040.00 DKK / 25µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a kinetochore protein that functions as part of the minichromosome instability-12 centromere complex. The encoded protein is required for proper kinetochore assembly and progression through the cell cycle. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: MTSVTRSEIIDEKGPVMSKTHDHQLESSLSPVEVFAKTSASLEMNQGVSEERIHLGSSPKKGGNCDLSHQERLQSKSLHLSPQEQSASYQDRRQSWRRASMKETNRRKSLHPIHQGITELSRSISVDLAESKRLGCLLLSSFQFSIQKLEPFLRDTKGFSLESFRAKASSLSEELKHFADGLETDGTLQKCFEDSNGKASDFSLEASVAEMKEYITKFSLERQTWDQLLLHYQQEAKEILSRGSTEAKITEVKVEPMTYLGSSQNEVLNTKPDYQKILQNQSKVFDCMELVMDELQGSVKQLQAFMDESTQCFQKVSVQLGKRSMQQLDPSPARKLLKLQLQNPPAIHGSGSGSCQ
Specifications

Specifications

AAH58899.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
66.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C20orf172/FLJ13346/MGC32987/MIS13/dJ469A13.2
DSN1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
79980
DSN1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTSVTRSEIIDEKGPVMSKTHDHQLESSLSPVEVFAKTSASLEMNQGVSEERIHLGSSPKKGGNCDLSHQERLQSKSLHLSPQEQSASYQDRRQSWRRASMKETNRRKSLHPIHQGITELSRSISVDLAESKRLGCLLLSSFQFSIQKLEPFLRDTKGFSLESFRAKASSLSEELKHFADGLETDGTLQKCFEDSNGKASDFSLEASVAEMKEYITKFSLERQTWDQLLLHYQQEAKEILSRGSTEAKITEVKVEPMTYLGSSQNEVLNTKPDYQKILQNQSKVFDCMELVMDELQGSVKQLQAFMDESTQCFQKVSVQLGKRSMQQLDPSPARKLLKLQLQNPPAIHGSGSGSCQ
RUO
DSN1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers