missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Dopa Decarboxylase/DDC Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB3564
This item is not returnable.
View return policy
Description
Dopa Decarboxylase/DDC Monoclonal specifically detects Dopa Decarboxylase/DDC in Human samples. It is validated for Western Blot, Knockout Validated.
Specifications
Dopa Decarboxylase/DDC | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Knockout Validated | |
P20711 | |
DDC | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human DDC, Met1-Glu480, Accession # P20711 | |
100 ÎĽg | |
Primary | |
Detects human DDC in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot, Immunoassay | |
413911 | |
Unconjugated | |
Lyophilized from a 0.2 ÎĽm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 ÎĽm filtered solution in PBS. with No Preservative | |
AADC, AADCaromatic-L-amino-acid decarboxylase, DDC, DOPA decarboxylase, dopa decarboxylase (aromatic L-amino acid decarboxylase), EC 4.1.1, EC 4.1.1.28 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1644 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human Dopa Decarboxylase/DDC Antibody, R&D Systems™ > 100ug; Unlabeled
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur