missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DIRAS1 Partial ORF (NP_660156.1, 99 a.a. - 198 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00148252-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16132077

  • 2735.00 DKK / 10µg
Estimated Shipment: 22-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

DIRAS1 belongs to a distinct branch of the functionally diverse Ras (see HRAS; MIM 190020) superfamily of monomeric GTPases.[supplied by OMIM]

Sequence: PIYKLIVQIKGSVEDIPVMLVGNKCDETQREVDTREAQAVAQEWKCAFMETSAKMNYNVKELFQELLTLETRRNMSLNIDGKRSGKQKRTDRVKGKCTLM
Specifications

Specifications

NP_660156.1
Liquid
148252
DIRAS1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Di-Ras1/FLJ42681/GBTS1/RIG
DIRAS1
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PIYKLIVQIKGSVEDIPVMLVGNKCDETQREVDTREAQAVAQEWKCAFMETSAKMNYNVKELFQELLTLETRRNMSLNIDGKRSGKQKRTDRVKGKCTLM
RUO
DIRAS1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers