missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DIP2A Partial ORF (NP_055966, 107 a.a. - 301 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00023181-Q03.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16075315

  • 4040.00 DKK / 25µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene may be involved in axon patterning in the central nervous system. This gene is not highly expressed. Several transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: KYKERKMPMPSKRRSVLVHSSVETYTPPDTSSASEDEGSLRRPGRLTSTPLQSHSSVEPWLDRVIQGSSTSSSASSTSSHPGGRPTTAPSAAATPGAAATTALAGLEAHTHIDLHSAPPDVTTGLVEHSYFERPQVASVRSVPRGCSGSMLETADGVPVNSRVSSKIQQLLNTLKRPKRPPLKEFFVDDFEELLE
Specifications

Specifications

NP_055966
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
47.275kDa
12.5% SDS-PAGE Stained with Coomassie Blue
KYKERKMPMPSKRRSVLVHSSVETYTPPDTSSASEDEGSLRRPGRLTSTPLQSHSSVEPWLDRVIQGSSTSSSASSTSSHPGGRPTTAPSAAATPGAAATTALAGLEAHTHIDLHSAPPDVTTGLVEHSYFERPQVASVRSVPRGCSGSMLETADGVPVNSRVSSKIQQLLNTLKRPKRPPLKEFFVDDFEELLE
RUO
DIP2A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
23181
DIP2A (Human) Recombinant Protein (Q03)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C21orf106/DIP2
DIP2A
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers