Få mere at vide
R&D Systems™ Human Cytokine Panel A Luminex™ Performance Control
Mængde:
1 stk.
Beskrivelse
- For use as quantitative controls for the determination of human cytokine concentrations in biological fluids.
- Concentrations have been assigned using R&D Systems Luminex™ High Performance Assays.
- Controls are prepared in a buffered protein solution with preservatives. They contain recombinant human cytokines at low, medium and high concentrations. Controls are supplied lyophilized.
Tekniske data
Tekniske data
| Produkttype | ELISA Kit |
| Mængde | 1 stk. |
| Arter | Human |
| Bindende mål | Luminex Assay Controls |
| Til brug med (udstyr) | Polystyrene and magnetic panels |
| Indhold og opbevaring | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |