missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human Cytokeratin 8 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB3165
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Cytokeratin 8 Monoclonal specifically detects Cytokeratin 8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Tekniske data
| Cytokeratin 8 | |
| Monoclonal | |
| LYOPH | |
| Western Blot, Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| CARD2, CK8, CK-8, CYK8, Cytokeratin-8, EndoA, K2C8, K8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, KRT8, Type-II keratin Kb8 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3856 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| LP3K | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| KRT8 | |
| SV-40 transformed human neonatal keratinocyte extract | |
| 100 μg | |
| Primary | |
| Detects human Cytokeratin 8. | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion