missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Human CXCR4 Synthetic Peptide

Artikelnummer. 15246229
Klik for at se tilgængelige muligheder
Mængde:
50 μg
Pakningsstørrelse:
50µg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 15246229

Brand: Invitrogen™ PEP0002

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Synthetic Peptide

CXCR4 Synthetic Peptide, PEP-0002, from Invitrogen.

CXCR4 (Fusin, LESTR, HUMSTR) is a member of the G-protein-coupled chemokine receptor family with seven membrane-spanning domains, and functions as a co-receptor for X4 HIV-1 entry into CD4+ cells. CXCR4 has been proved to be the co-receptor for HIV-2's binding to CD4 through envelope glycoprotein gp 120 and promotes Env-mediated fusion of the virus. In other cases, CXCR4 can even function as the only receptor for HIV-2's binding to the CD4 host cells. CXCR4 is the receptor for the C-X-C chemokine CXCL12/SDF-1 that transduces a signal by increasing intracellular calcium ion levels and enhancing MAPK1/MAPK3 activation. CXCR4 acts as a receptor for extracellular ubiquitin; leading to enhanced intracellular calcium ions and reduced cellular cAMP levels. Also, CXCR4 is involved in hematopoiesis and in cardiac ventricular septum formation, and plays an essential role in vascularization of the gastrointestinal tract, probably by regulating vascular branching and/or remodeling processes in endothelial cells. In the CNS, CXCR4 could mediate hippocampal-neuron survival and is involved in cerebellar development. CXCR4 binds bacterial lipopolysaccharide (LPS) and mediates LPS-induced inflammatory response, including TNF secretion by monocytes. Antibodies to CXCR4 block HIV-1 and HIV-2 fusion and infection of human target cells. The amino-terminal domain and the second extracellular loop of CXCR4 serve as HIV biding sites.
TRUSTED_SUSTAINABILITY

Tekniske data

Gen-id (Entrez) 7852
Arter Human
Oprensningsmetode purified
Adgangsnummer P61073
Fælles navn Human CXCR4
Koncentration 200 μg/mL
Indhold og opbevaring -20°C
Oversigt Synthetic Peptide
Format Liquid
Formulering PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
Til brug med (applikation) Blocking Assay, Control
Gen symbol CXCR4
Lyofiliseret, frysetørret No
Konserveringsmiddel 0.02% sodium azide
Produkttype Peptide
Mængde 50 μg
Regulatorisk status RUO - research use only
Opbevaringsbuffer PBS
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.