Learn More
Abcam Human CXCL10 (IP10) knockout THP-1 cell line
Product Code. 32157801 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1,000,000 Cells
2 x 1,000,000 Cells
Unit Size:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32157801 2 x 1,000,000 Cells Pack of 2
32155979 1,000,000 Cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 32157801 Supplier Abcam Supplier No. ab2778602x1000000Cells_
 Restricted Item
Forventet forsendelse: 27-11-2026
CXCL10 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 80 bp Deletion in Exon 2. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knocko...
CXCL10 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 80 bp Deletion in Exon 2. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells
Regulatory Status RUO