missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Gibco™ Human CXCL1 (GRO alpha) Recombinant Protein, PeproTech®
Klik for at se tilgængelige muligheder
Mængde:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
Beskrivelse
Recombinant Human GRO-alpha/MGSA is a 7.8 kDa protein consisting of 73 amino acids, including the 'ELR' motif common to the CXC chemokine family that binds to CXCR1 or CXCR2. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
CXCL1, CXCL2, and CXCL3, also known respectively as GRO alpha, GRO beta (MIP-2 alpha) and GRO gamma (MIP-2 beta), are members of the CXC subfamily of chemokines. Mature CXCL1/2/3 proteins bind with high affinity to the IL-8 receptor type B and are potent neutrophil attractants and activators. In vitro, the processed forms GRO-alpha(4-73), GRO-alpha(5-73) and GRO-alpha(6-73) show a 30-fold higher chemotactic activity.
Tekniske data
Tekniske data
| Adgangsnummer | P09341 |
| Til brug med (applikation) | ELISA, Functional Assay, Western Blot |
| Formulering | protein with no preservative |
| Gen-id (Entrez) | 2919 |
| Molekylvægt (g/mol) | 7.8 kDa |
| Navn | Human CXCL1 (GRO alpha) |
| Mængde | 100 μg |
| Kilde | E. coli |
| Regulatorisk status | RUO |
| Endotoksin koncentration | <1 EU/ μg |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion