Learn More
Abnova™ Human CTDP1 Partial ORF (AAH63447.1, 332 a.a. - 431 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009150-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein which interacts with the carboxy-terminus of transcription initiation factor TFIIF, a transcription factor which regulates elongation as well as initiation by RNA polymerase II. The protein may also represent a component of an RNA polymerase II holoenzyme complex. Alternative splicing of this gene results in two transcript variants encoding 2 different isoforms. [provided by RefSeq]
Sequence: PPGSRESQTRKKVNHSRGTEVSEPSPPVRDPEGVTQAPGVEPSNGLEKPARELNGSEAATPRDSPRPGKPDERDIWPPAQAPTSSQELAGAPEPQGSCAQSpecifications
AAH63447.1 | |
Liquid | |
9150 | |
CTDP1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CCFDN/FCP1 | |
CTDP1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PPGSRESQTRKKVNHSRGTEVSEPSPPVRDPEGVTQAPGVEPSNGLEKPARELNGSEAATPRDSPRPGKPDERDIWPPAQAPTSSQELAGAPEPQGSCAQ | |
RUO | |
CTDP1 | |
Wheat Germ (in vitro) | |
GST |