missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Coronavirus Spike Protein Mouse anti-HCoV-229E, Clone: 11, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14653-100UL
Additional Details : Weight : 0.00970kg
Description
Human Coronavirus Spike Protein Monoclonal antibody specifically detects Human Coronavirus Spike Protein in HCoV-229E samples. It is validated for Western Blot, ELISASpecifications
Human Coronavirus Spike Protein | |
Monoclonal | |
Unconjugated | |
0.2μm filtered solution in PBS without Preservative | |
Mouse | |
Protein A purified | |
RUO | |
3200426 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA | |
11 | |
Western Blot 1:1000-1:5000, ELISA 1:1000-1:2000 | |
HKU1, Spike glycoprotein | |
This product is a recombinant monoclonal antibody expressed from HEK293 cells. | |
100 μg | |
Primary | |
HCoV-229E | |
Purified |