missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human Collagen XIII alpha 1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6346-SP
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Collagen XIII alpha 1 Polyclonal specifically detects Collagen XIII alpha 1 in Human samples. It is validated for Western Blot.
Tekniske data
| Collagen XIII alpha 1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| COL13A1, collagen alpha-1(XIII) chain, collagen, type XIII, alpha 1, FLJ42485 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1305 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| NP_542992 | |
| COL13A1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Collagen XIII Glu109-Leu668 Accession # NP_542992 | |
| 25 μg | |
| Primary | |
| Detects human Collagen XIII in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse COL13A1 and less than 1% cross-reactivity with recombinant human (rh) COL23A1 and rhCOL25A1 is observed. | |
| Human | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion