Learn More
Abnova™ Human COL5A2 Partial ORF (NP_000384, 41 a.a. - 124 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001290-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes an alpha chain for one of the low abundance fibrillar collagens. Fibrillar collagen molecules are trimers that can be composed of one or more types of alpha chains. Type V collagen is found in tissues containing type I collagen and appears to regulate the assembly of heterotypic fibers composed of both type I and type V collagen. This gene product is closely related to type XI collagen and it is possible that the collagen chains of types V and XI constitute a single collagen type with tissue-specific chain combinations. Mutations in this gene are associated with Ehlers-Danlos syndrome, types I and II. [provided by RefSeq]
Sequence: CTQNGQMYLNRDIWKPAPCQICVCDNGAILCDKIECQDVLDCADPVTPPGECCPVCSQTPGGGNTNFGRGRKGQKGEPGLVPVVSpecifications
NP_000384 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.98kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
CTQNGQMYLNRDIWKPAPCQICVCDNGAILCDKIECQDVLDCADPVTPPGECCPVCSQTPGGGNTNFGRGRKGQKGEPGLVPVV | |
RUO | |
COL5A2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1290 | |
COL5A2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC105115 | |
COL5A2 | |
Recombinant | |
wheat germ expression system |