Learn More
Abnova™ Human CHMP1B Partial ORF (NP_065145, 145 a.a. - 199 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00057132-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
CHMP1B belongs to the chromatin-modifying protein/charged multivesicular body protein (CHMP) family. These proteins are components of ESCRT-III (endosomal sorting complex required for transport III), a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies (MVBs). Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A (MIM 164010), is required for both MVB formation and regulation of cell cycle progression (Tsang et al., 2006 [PubMed 16730941]).[supplied by OMIM]
Sequence: PQNQVDMLLQEMADEAGLDLNMELPQGQTGSVGTSVASAEQDELSQRLARLRDQVSpecifications
NP_065145 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.79kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PQNQVDMLLQEMADEAGLDLNMELPQGQTGSVGTSVASAEQDELSQRLARLRDQV | |
RUO | |
CHMP1B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
57132 | |
CHMP1B (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C10orf2/C18-ORF2/C18orf2/CHMP1.5/Vps46-2 | |
CHMP1B | |
Recombinant | |
wheat germ expression system |