missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CH25H Partial ORF (AAH17843.1, 142 a.a. - 247 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009023-Q03.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16085115

  • 4040.00 DKK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This is an intronless gene that is involved in cholesterol and lipid metabolism. The encoded protein is a membrane protein and contains clusters of histidine residues essential for catalytic activity. Unlike most other sterol hydroxylases, this enzyme is a member of a small family of enzymes that utilize diiron cofactors to catalyze the hydroxylation of hydrophobic substrates. [provided by RefSeq]

Sequence: WHLLHHKVPWLYRTFHKVHHQNSSSFALATQYMSVWELFSLGFFDMMNVTLLGCHPLTTLTFHVVNIWLSVEDHSGYNFPWSTHRLVPFGWYGGVVHHDLHHSHFN
Specifications

Specifications

AAH17843.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.29kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
WHLLHHKVPWLYRTFHKVHHQNSSSFALATQYMSVWELFSLGFFDMMNVTLLGCHPLTTLTFHVVNIWLSVEDHSGYNFPWSTHRLVPFGWYGGVVHHDLHHSHFN
RUO
CH25H
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9023
CH25H (Human) Recombinant Protein (Q03)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C25H
CH25H
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers