missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CFP Partial ORF (AAH15756.1, 213 a.a. - 334 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005199-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16178941

  • 4040.00 DKK / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a plasma glycoprotein that positively regulates the alternative complement pathway of the innate immune system. This protein binds to many microbial surfaces and apoptotic cells and stabilizes the C3- and C5-convertase enzyme complexes in a feedback loop that ultimately leads to formation of the membrane attack complex and lysis of the target cell. Mutations in this gene result in two forms of properdin deficiency, which results in high susceptibility to meningococcal infections. Multiple alternatively spliced variants, encoding the same protein, have been identified

Sequence: PHEPKETRSRKCSAPEPSQKPPGKPCPGLAYEQRRCTGLPPCPVAGGWGPWGPVSPCPVTCGLGQTMEQRTCNHPVPQHGGPFCAGDATRTHICNTAVPCPVDGEWDSWGEWSPCIRRNMKS
Specifications

Specifications

AAH15756.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
39.16kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PHEPKETRSRKCSAPEPSQKPPGKPCPGLAYEQRRCTGLPPCPVAGGWGPWGPVSPCPVTCGLGQTMEQRTCNHPVPQHGGPFCAGDATRTHICNTAVPCPVDGEWDSWGEWSPCIRRNMKS
RUO
CFP
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5199
CFP (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BFD/PFC/PFD/PROPERDIN
CFP
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers