Learn More
Human CFHR3 (Complement factor H-related protein 3) knockout A549 cell lysate, 1 Kit
Product Code. 32148405 Shop All Abcam Products
Product Code. 32148405 Supplier Abcam Supplier No. ab2631481Kit
 Restricted Item
Forventet forsendelse: 07-12-2026
Human CFHR3 (Complement factor H-related protein 3) knockout A549 cell/tissue lysate.
  • Species: Human
  • Tissue: CFHR3 (Complement factor H-related protein 3) knockout A549