missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CD38 His-tag, Recombinant Protein
HEK293 cells Recombinant Protein
890.00 DKK - 11560.00 DKK
Specifications
Accession Number | P28907 |
---|---|
For Use With (Application) | Control, ELISA, Immunohistochemistry, Western Blot |
Formulation | protein with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 |
Gene ID (Entrez) | 952 |
Name | Human CD38 His-tag |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
17174233
|
Invitrogen™
A42569 |
5 μg |
890.00 DKK
5µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17184233
|
Invitrogen™
A42570 |
50 μg |
3150.00 DKK
50µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17154233
|
Invitrogen™
A42567 |
5 x 5 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17164233
|
Invitrogen™
A42568 |
5 x 50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Beschreibung
CD38 (NAD+ glycohydrolase) is a type II transmembrane glycoprotein able to induce activation, proliferation and differentiation of mature lymphocytes and mediate apoptosis of myeloid and lymphoid progenitor cells. CD38 functions as a multi-catalytic ectoenzyme serving as ADP-ribosyl cyclase, cyclic ADP-ribose hydrolase and possibly NAD+ glycohydrolase or as a cell surface receptor. Antibodies to CD38 are useful in subtyping of lymphomas and leukemias, detection of plasma cells (i.e. identification of myelomas), and as a marker for activated B and T cells. CD38 participates in cell adhesion, signal transduction and calcium signaling. Further, CD38 is expressed at high levels in the pancreas, liver, kidney, malignant lymphoma and neuroblastoma. Diseases associated with CD38 dysfunction include chronic lymphocytic leukemia and Richter's Syndrome.Spezifikation
P28907 | |
protein with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | |
Human CD38 His-tag | |
-20°C | |
<1 EU/μg | |
CD38 | |
Unconjugated | |
Lyophilized |
Control, ELISA, Immunohistochemistry, Western Blot | |
952 | |
HEK293 cells | |
RUO | |
2 '-phospho-ADP-ribosyl cyclase; 2 '-phospho-ADP-ribosyl cyclase/2 '-phospho-cyclic-ADP-ribose transferase; 2 '-phospho-cyclic-ADP-ribose transferase; ADPRC 1; ADPRC1; ADP-ribosyl cyclase 1; ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 1; cADPr hydrolase 1; Cd38; CD38 antigen; CD38 antigen (ADP-ribosyl cyclase / cyclic ADP-ribose hydrolase); CD38 antigen (p45); CD38 antigen p45; CD38 molecule; CD38H; Cd38-rs1; cluster of differentiation 38; cyclic ADP-ribose hydrolase 1; ecto-nicotinamide adenine dinucleotide glycohydrolase; I-19; NAD(+) nucleosidase; NAD+ nucleosidase; NIM-R5 antigen; T10 | |
Human | |
Recombinant | |
≥ 90% |