missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human CD31/PECAM-1 Antibody, Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB11666-100
3462.33 DKK valid until 2025-12-16
BEDSTE tilbudspris! Brug kampagnekoden "24090" for at få din tilbudspris.
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CD31/PECAM-1 Monoclonal antibody specifically detects CD31/PECAM-1 in Human samples. It is validated for Western Blot,Simple Western,Immunohistochemistry,Multiplex Immunofluorescence
Tekniske data
| CD31/PECAM-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
| adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
| Chinese Hamster Ovary cell line, CHO-derived recombinant human CD31/PECAM01, Gln28-Lys601, Accession # P16284 | |
| 100 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| Purified |
| Western Blot, Simple Western, Immunohistochemistry, Multiplex Immunofluorescence | |
| 1094431 | |
| Western Blot 2 μg/mL, Simple Western 5 μg/mL, Immunohistochemistry 0.05-10 μg/mL, Multiplex Immunofluorescence 0.1 μg/mL | |
| P16284 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5175 | |
| Human | |
| Antibody | |
| IgG2b |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion