Få mere at vide
Human CD31/PECAM-1 Antibody, Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB11666-SP
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CD31/PECAM-1 Monoclonal antibody specifically detects CD31/PECAM-1 in Human samples. It is validated for Western Blot,Simple Western,Immunohistochemistry,Multiplex Immunofluorescence
Tekniske data
| CD31/PECAM-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
| adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
| Chinese Hamster Ovary cell line, CHO-derived recombinant human CD31/PECAM01, Gln28-Lys601, Accession # P16284 | |
| 25 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| Purified |
| Western Blot, Simple Western, Immunohistochemistry, Multiplex Immunofluorescence | |
| 1094431 | |
| Western Blot 2 μg/mL, Simple Western 5 μg/mL, Immunohistochemistry 0.05-10 μg/mL, Multiplex Immunofluorescence 0.1 μg/mL | |
| P16284 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5175 | |
| Human | |
| Antibody | |
| IgG2b |