missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human CD19 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB4867T-025
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CD19 Monoclonal specifically detects CD19 in Human samples. It is validated for Flow Cytometry.
Tekniske data
| CD19 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD19 | |
| Human CLL cells | |
| 25 Tests | |
| Primary | |
| Detects human CD19. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 4G7-2E3 | |
| Flow Cytometry 5 uL/10^6 cells | |
| B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, Leu-12, MGC12802, T-cell surface antigen Leu-12 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 930 | |
| Human | |
| Supernatant |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion