Learn More
Abnova™ Human CD164 Full-length ORF (AAH11522.1, 1 a.a. - 197 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008763-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Sialomucins are a heterogeneous group of secreted or membrane-associated mucins that appear to play 2 key but opposing roles in vivo: first as cytoprotective or antiadhesive agents, and second as adhesion receptors. CD164 is a type I integral transmembrane sialomucin that functions as an adhesion receptor (Watt et al., 1998 [PubMed 9680353]; Forde et al., 2007 [PubMed 17077324]).[supplied by OMIM]
Sequence: MSRLSRSLLWAATCLGVLCVLSADKNTTQHPNVTTLAPISNVTSAPVTSLPLVTTPAPETCEGRNSCVSCFNVSVVNTTCFWIECKDESYCSHNSTVSDCQVGNTTDFCSVSTATPVPTANSTAKPTVQPSPSTTSKTVTTSGTTNNTVTPTSQPVRKSTFDAASFIGGIVLVLGVQAVIFFLYKFCKSKERNYHTLSpécification
AAH11522.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
47.41kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC-24/MUC-24/endolyn | |
CD164 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8763 | |
CD164 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSRLSRSLLWAATCLGVLCVLSADKNTTQHPNVTTLAPISNVTSAPVTSLPLVTTPAPETCEGRNSCVSCFNVSVVNTTCFWIECKDESYCSHNSTVSDCQVGNTTDFCSVSTATPVPTANSTAKPTVQPSPSTTSKTVTTSGTTNNTVTPTSQPVRKSTFDAASFIGGIVLVLGVQAVIFFLYKFCKSKERNYHTL | |
RUO | |
CD164 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |