missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human CD11b/Integrin alpha M Biotinylated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems BAM1699
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CD11b Monoclonal specifically detects CD11b in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.
Tekniske data
| CD11b | |
| Monoclonal | |
| Biotin | |
| Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
| ITGAM | |
| Mouse B cells | |
| 100 μg | |
| Primary | |
| Detects human Integrin alpha M/CD11b in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry | |
| ICRF44 | |
| Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL | |
| antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, Integrin alpha M, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), ITGAM, Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3684 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion