missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human CD-M6PR Lumenal Domain Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5320
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CD-M6PR Polyclonal specifically detects CD-M6PR in Human samples. It is validated for Western Blot.
Tekniske data
| CD-M6PR | |
| Polyclonal | |
| Unconjugated | |
| P20645 | |
| M6PR | |
| Mouse myeloma cell line NS0-derived recombinant human CD-M6PR Lumenal Domain Thr27-His185 Accession # P20645 | |
| 100 μg | |
| Primary | |
| Detects human CD-M6PR Lumenal Domain in direct ELISAs and Western blots. In direct ELISAs, approximately 60% cross-reactivity with recombinant mouse CD-M6PR is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| 000 Man6PR, 46 kDa mannose 6-phosphate receptor, 46-kDa mannose 6-phosphate receptor, cation-dependent mannose-6-phosphate receptor, CD Man-6-P receptor, CDM6PR, CD-MPR, FLJ32994, M6PR, mannose-6-phosphate receptor (cation dependent), MPR 46, MPR46, MPRD, Mr 46, SMPR | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4074 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion