missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CCL2 (MCP-1) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP1548
Additional Details : Weight : 0.01000kg
Description
CCL2 Synthetic Peptide, PEP-1548, from Invitrogen.
This peptide corresponds to 17 amino acids near the carboxy terminus of human CCL2.PEP-1548 can be used as a blocking peptide with polyclonal antibody PA5-34505.Specifications
6347 | |
purified | |
Human CCL2 (MCP-1) | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
P13500 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
CCL2 | |
0.02% sodium azide | |
50 μg | |
PBS |