missing translation for 'onlineSavingsMsg'
Få mere at vide

Gibco™ Human CCL11 (Eotaxin), Animal-Free Recombinant Protein, PeproTech®

Artikelnummer. 17843063
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
100 μg
20 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17843063 100 μg
17823233 1 mg
17823693 20 μg
17823623 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17843063 Leverandør Gibco™ Leverandørnr. AF30021100UG

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Recombinant Protein

Recombinant Human Eotaxin is an 8.3 kDa protein containing 74 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene is one of several Cys-Cys (CC) cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil specific chemokine assumed to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.
TRUSTED_SUSTAINABILITY

Tekniske data

Adgangsnummer P51671
Til brug med (applikation) Functional Assay
Formulering protein with no preservative
Gen-id (Entrez) 6356
Molekylvægt (g/mol) 8.3 kDa
Navn Human CCL11 (Eotaxin), Animal-Free
Mængde 100 μg
Kilde E. coli
Regulatorisk status RUO
Endotoksin koncentration <0.1 EU/ μg
Gene Alias C-C motif chemokine 11; C-C motif chemokine ligand 11; Ccl11; chemokine (C-C motif) ligand 11; eosinophil chemotactic protein; Eotaxin; Eotaxin 1; Eotaxin1; eotaxin-1; MGC22554; Scya11; small chemokine (C-C motif) ligand 11; small inducible cytokine A11; small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin); small inducible cytokine subfamily A11; small-inducible cytokine A11
Fælles navn Eotaxin
Gen symbol CCL11
Konjugeret Unconjugated
Rekombinant Recombinant
Sekvens GPASVPTTCC FNLANRKIPL QRLESYRRIT SGKCPQKAVI FKTKLAKDIC ADPKKKWVQD SMKYLDQKSP TPKP
Indhold og opbevaring -20°C
Udtrykssystem E. coli
Form Lyophilized
Renhed eller kvalitetsklasse ≥ 98% by SDS-PAGE gel and HPLC analyses.
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.