missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Human Caspase 13 Synthetic Peptide

Artikelnummer. 15246299
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 μg
Pakningsstørrelse:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15246299 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15246299 Leverandør Invitrogen™ Leverandørnr. PEP0074

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Synthetic Peptide

This peptide corresponds to 15 amino acids of human Caspase-13. PEP-0074 can be used as a blocking peptide with polyclonal antibody PA5-19949.

Apoptosis is related to many diseases and induced by a family of cell death receptors and their ligands. Cell death is finally caused by members of the caspase family of proteases and caspase activated DNases. A novel member in the caspase family was recently identified and designated ERICE (for Evolutionarily Related Interleukin-1 beta Converting Enzyme) and caspase-13. Caspase-13 belongs to the ICE subfamily of caspases. Overexpression of caspase-13 induces apoptosis. Caspase-13 was activated by caspase-8, which is a key enzyme in death receptor induced apoptosis. Caspase-13 is expressed in a variety of human tissues and cell lines.
TRUSTED_SUSTAINABILITY

Tekniske data

Gen-id (Entrez) 8628
Arter Human
Oprensningsmetode purified
Adgangsnummer P49662
Fælles navn Caspase 13
Koncentration 200 μg/mL
Indhold og opbevaring -20°C
Oversigt Synthetic Peptide
Format Liquid
Formulering PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
Til brug med (applikation) Blocking Assay, Control
Gene Alias apoptosis-related cysteine protease; Casp11; CASP-11; CASP13; CASP-13; CASP4; CASP-4; caspase 11; caspase 4; caspase 4, apoptosis-related cysteine peptidase; caspase 4, apoptosis-related cysteine protease; caspase 4, isoform alpha, small subunit; Caspase-11; Caspase-13; Caspase-13 subunit 1; Caspase-13 subunit 2; caspase-13A; caspase-13B; caspase-4; Caspase-4 subunit 1; Caspase-4 subunit 2; Caspase-4 subunit p10; Caspase-4 subunit p20; Caspl; ERICE; Evolutionary related interleukin-1-beta-converting enzyme; Ich3; ich-3; MGC137679; Protease ICH-3
Gen symbol CASP13
Lyofiliseret, frysetørret No
Konserveringsmiddel Sodium azide
Produkttype Peptide
Mængde 50 μg
Regulatorisk status RUO
Opbevaringsbuffer PBS
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.