Learn More
Abnova™ Human CARD14 Partial ORF (NP_077015, 905 a.a. - 1004 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_077015 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 79092 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16075645
|
Abnova™
H00079092-Q02.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16065645
|
Abnova™
H00079092-Q02.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene belongs to the membrane-associated guanylate kinase (MAGUK) family, a class of proteins that functions as molecular scaffolds for the assembly of multiprotein complexes at specialized regions of the plasma membrane. This protein is also a member of the CARD protein family, which is defined by carrying a characteristic caspase-associated recruitment domain (CARD). This protein shares a similar domain structure with CARD11 protein. The CARD domains of both proteins have been shown to specifically interact with BCL10, a protein known to function as a positive regulator of cell apoptosis and NF-kappaB activation. When expressed in cells, this protein activated NF-kappaB and induced the phosphorylation of BCL10. Two alternatively spliced variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq]
Sequence: VQLDSVCTLHRMDIFPIVIHVSVNEKMAKKLKKGLQRLGTSEEQLLEAARQEEGDLDRAPCLYSSLAPDGWSDLDGLLSCVRQAIADEQKKVVWTEQSPRSpecifications
NP_077015 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BIMP2/CARMA2 | |
CARD14 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
79092 | |
CARD14 (Human) Recombinant Protein (Q02) | |
VQLDSVCTLHRMDIFPIVIHVSVNEKMAKKLKKGLQRLGTSEEQLLEAARQEEGDLDRAPCLYSSLAPDGWSDLDGLLSCVRQAIADEQKKVVWTEQSPR | |
RUO | |
CARD14 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |