missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Human Carbonic Anhydrase II (aa 7-144) Control Fragment Recombinant Protein

Artikelnummer. 30206910
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30206910 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30206910 Leverandør Invitrogen™ Leverandørnr. RP95151

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51598 (PA5-51598. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

CA2 is one of several (at least 7) isozymes of carbonic anhydrase. Carbonic anhydrase catalyzes reversible hydration of carbon dioxide. Defects in this enzyme are associated with osteopetrosis and renal tubular acidosis.
TRUSTED_SUSTAINABILITY

Tekniske data

Adgangsnummer P00918
Koncentration ≥5.0 mg/mL
Til brug med (applikation) Blocking Assay, Control
Formulering 1 M urea, PBS with no preservative; pH 7.4
Gen-id (Entrez) 760
Navn Human Carbonic Anhydrase II (aa 7-144) Control Fragment
Mængde 100 μL
Regulatorisk status RUO
Gene Alias AI131712; CA II; CA2; CAC; CAII; CA-II; Car2; Car-2; Carbonate dehydratase II; carbonic anhydrase 2; carbonic anhydrase B; carbonic anhydrase C; Carbonic anhydrase II; carbonic dehydratase; epididymis luminal protein 76; epididymis secretory protein Li 282; HEL-76; HEL-S-282; Ltw-5; Lvtw-5
Fælles navn Carbonic Anhydrase II
Gen symbol CA2
Konjugeret Unconjugated
Arter Human
Rekombinant Recombinant
Protein tag His-ABP-tag
Sekvens YGKHNGPEHWHKDFPIAKGERQSPVDIDTHTAKYDPSLKPLSVSYDQATSLRILNNGHAFNVEFDDSQDKAVLKGGPLDGTYRLIQFHFHWGSLDGQGSEHTVDKKKYAAELHLVHWNTKYGDFGKAVQQPDGLAVLG
Indhold og opbevaring -20°C, Avoid Freeze/Thaw Cycles
Udtrykssystem E. coli
Form Liquid
Renhed eller kvalitetsklasse >80% by SDS-PAGE and Coomassie blue staining
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.