missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CAPN11 Partial ORF (NP_008989, 557 a.a. - 651 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00011131-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16136222

  • 4150.00 DKK / 25µg
Estimated Shipment: 10-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Calpains constitute a family of intracellular calcium-dependent cysteine proteases. There are eight members in this superfamily. They consist of a variable 80 kDa subunit and an invariant 30 kDa subunit. This calpain protein appears to have protease activity and calcium-binding ability. A similar mouse protein may play a functional role in spermatogenesis and in the regulation of calcium-dependent signal transduction events during meiosis. [provided by RefSeq]

Sequence: LLNRMAIKFKSFKTKGFGLDACRCMINLMDKDGSGKLGLLEFKILWKKLKKWMDIFRECDQDHSGTLNSYEMRLVIEKAGIKLNNKVMQVLVARY
Specifications

Specifications

NP_008989
Liquid
11131
CAPN11 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
calpain11
CAPN11
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.19kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LLNRMAIKFKSFKTKGFGLDACRCMINLMDKDGSGKLGLLEFKILWKKLKKWMDIFRECDQDHSGTLNSYEMRLVIEKAGIKLNNKVMQVLVARY
RUO
CAPN11
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers