missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human/Canine CD11b/Integrin alpha M Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1699
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CD11b Polyclonal specifically detects CD11b in Human, Canine samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Tekniske data
| CD11b | |
| Polyclonal | |
| Unconjugated | |
| NP_001139280 | |
| ITGAM | |
| Mouse myeloma cell line NS0-derived recombinant human CD11b/Integrin alpha M Phe17-Asn1105 Accession # NP_001139280 | |
| 100 μg | |
| Primary | |
| Detects human CD11b/Integrin alpha M in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse Integrin alpha M and recombinant human Integrin alpha E is observed. | |
| Human, Canine | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, Integrin alpha M, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), ITGAM, Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3684 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion