missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human C7orf11 Partial ORF (NP_619646.1, 80 a.a. - 179 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00136647-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16131877

  • 2735.00 DKK / 10µg
Estimated Shipment: 22-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: SPSPGGYPGSYSRSPAGSQQQFGYSPGQQQTHPQGSPRTSTPFGSGRVREKRMSNELENYFKPSMLEDPWAGLEPVSVVDISQQYSNTQTFTGKKGRYFC
Specifications

Specifications

NP_619646.1
Liquid
136647
C7orf11 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ABHS/ORF20/TTDN1
C7orf11
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SPSPGGYPGSYSRSPAGSQQQFGYSPGQQQTHPQGSPRTSTPFGSGRVREKRMSNELENYFKPSMLEDPWAGLEPVSVVDISQQYSNTQTFTGKKGRYFC
RUO
C7orf11
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers