missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human C21orf66 Full-length ORF (AAH30539.1, 1 a.a. - 398 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | AAH30539.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 94104 |
Molecular Weight (g/mol) | 70.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16136883
|
Abnova™
H00094104-P01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16126883
|
Abnova™
H00094104-P01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein that may bind to GC-rich DNA sequences, which suggests its involvement in the regulation of transcription. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq]
Sequence: MFRKARRVNVRKRNDSEEEERERDEEQEPPPLLPPPGTGEEAGPGGGDRAPGGESLLGPGPSPPSALTPGLGAEAGGGFPGGAEPGNGLKPRKRPRENKEVPRASLLSFQDEEEENEEVFKVKKSSYSKKIVKLLKKEYKEDLEKSKIKTELNSSAESEQPLDKTGHVKDTNQEDGVIISEHGEDEMDMESEKEEEKPKTGGAFSNALSSLNVLRPGEIPDAAFIHAARKKRQMARELGDFTPHDNEPGKGRLVREDENDASDDEDDDEKRRIVFSVKEKSQRQKIAEEIGIEGSDDDALVTGEQDEELSRWEQEQIRKGINIPQVQASQPAEVNMYYQNTYQTMPYGSSYGIPYSYTAYGSSDAKSQKTDNTVPFKTPSNEMTPVTIDLVKKQLKDRSpecifications
AAH30539.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
70.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MFRKARRVNVRKRNDSEEEERERDEEQEPPPLLPPPGTGEEAGPGGGDRAPGGESLLGPGPSPPSALTPGLGAEAGGGFPGGAEPGNGLKPRKRPRENKEVPRASLLSFQDEEEENEEVFKVKKSSYSKKIVKLLKKEYKEDLEKSKIKTELNSSAESEQPLDKTGHVKDTNQEDGVIISEHGEDEMDMESEKEEEKPKTGGAFSNALSSLNVLRPGEIPDAAFIHAARKKRQMARELGDFTPHDNEPGKGRLVREDENDASDDEDDDEKRRIVFSVKEKSQRQKIAEEIGIEGSDDDALVTGEQDEELSRWEQEQIRKGINIPQVQASQPAEVNMYYQNTYQTMPYGSSYGIPYSYTAYGSSDAKSQKTDNTVPFKTPSNEMTPVTIDLVKKQLKDR | |
RUO | |
C21orf66 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
94104 | |
C21orf66 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BM-020/FLJ90561/GCFC/fSAP105 | |
C21orf66 | |
Recombinant | |
wheat germ expression system |