missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Human C15orf40 (aa 87-146) Control Fragment Recombinant Protein
Klik for at se tilgængelige muligheder
Mængde:
100 μL
Beskrivelse
Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60184 (PA5-60184. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
C15orf40 is a protein coding gene.
Tekniske data
Tekniske data
| Adgangsnummer | Q8WUR7 |
| Koncentration | ≥5.0 mg/mL |
| Til brug med (applikation) | Blocking Assay, Control |
| Formulering | 1 M urea, PBS with no preservative; pH 7.4 |
| Gen-id (Entrez) | 123207 |
| Navn | Human C15orf40 (aa 87-146) Control Fragment |
| Mængde | 100 μL |
| Regulatorisk status | RUO |
| Gene Alias | 3110040N11Rik; AI851475; C15orf40; chromosome 15 open reading frame 40; RIKEN cDNA 3110040N11 gene; UPF0235 protein C15orf40; UPF0235 protein C15orf40 homolog |
| Fælles navn | C15orf40 |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion