missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BRI3BP Full-length ORF (NP_542193.3, 1 a.a. - 251 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00140707-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138243

  • 4040.00 DKK / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MGARASGGPLARAGLLLLLLLLLLLGLLAPGAQGARGRGGAEKNSYRRTVNTFSQSVSSLFGEDNVRAAQKFLARLTERFVLGVDMFVETLWKVWTELLDVLGLDVSNLSQYFSPASVSSSPARALLLVGVVLLAYWFLSLTLGFTFSVLHVVFGRFFWIVRVVLFSMSCVYILHKYEGEPENAVLPLCFVVAVYFMTGPMGFYWRSSPSGPSNPSNPSVEEKLEHLEKQVRLLNIRLNRVLESLDRSKDK
Specifications

Specifications

NP_542193.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
54.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
BRI3BP
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
140707
BRI3BP (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BNAS1/HCCR-2/KG19
BRI3BP
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers