missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Human BPIFB4 Control Fragment Recombinant Protein
Klik for at se tilgængelige muligheder
Mængde:
100 μL
Beskrivelse
Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-144817 (PA5-144817. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Tekniske data
Tekniske data
| Adgangsnummer | P59827 |
| Koncentration | ≥5.0 mg/mL |
| Til brug med (applikation) | Blocking Assay, Control |
| Formulering | 1 M urea, PBS with no preservative; pH 7.4 |
| Gen-id (Entrez) | 149954 |
| Navn | Human BPIFB4 Control Fragment |
| Mængde | 100 μL |
| Regulatorisk status | RUO |
| Gene Alias | BPI fold containing family B member 4; BPI fold containing family B, member 4; BPI fold-containing family B member 4; BPIFB4; C20orf186; dJ726C3.5; Gm1006; LBP / BPI / CETP family, N-terminal domain containing protein; ligand-binding protein RY2G5; likely ortholog of rat probable ligand-binding protein RY2G5; long palate, lung and nasal epithelium carcinoma associated 4; long palate, lung and nasal epithelium carcinoma-associated protein 4; LPLUNC4; Novel LBP/BPI/CETP family, N-terminal domain containing protein; potential ligand-binding protein; RY2G5 |
| Fælles navn | BPIFB4 |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion