missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human BOLA1 Full-length ORF (NP_057158.1, 1 a.a. - 137 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051027-P01.10ug
This item is not returnable.
View return policy
Description
Sequence: MLSGRLVLGLVSMAGRVCLCQGSAGSGAIGPVEAAIRTKLEEALSPEVLELRNESGGHAVPPGSETHFRVAVVSSRFEGLSPLQRHRLVHAALAEELGGPVHALAIQARTPAQWRENSQLDTSPPCLGGNKKTLGTPSpecifications
NP_057158.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
40.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ÎĽg | |
RUO | |
BOLA1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
51027 | |
BOLA1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CGI-143/MGC75015/RP11-196G18.18 | |
BOLA1 | |
Recombinant | |
wheat germ expression system |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ Human BOLA1 Full-length ORF (NP_057158.1, 1 a.a. - 137 a.a.) Recombinant Protein with GST-tag at N-terminal > 10μg
Spot an opportunity for improvement?Share a Content Correction