missing translation for 'onlineSavingsMsg'
Få mere at vide

Gibco™ Human BMP-4 Recombinant Protein, PeproTech®

Artikelnummer. 30088425
Klik for at se tilgængelige muligheder
Mængde:
1 μg
10 x 100 μg
100 μg
5 μg
5 x 100 μg
50 μg
Pakningsstørrelse:
5µg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 30088425

Brand: Gibco™ 1200501M

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Recombinant Protein

120-05-1MG will be provided as 10 x 100 μg (120-05-100UG). 120-05-500UG will be provided as 5 x 100 μg (120-05-100UG). Recombinant Human BMP-4, expressed in HeLa cells, is a 25.6 kDa homodimeric glycoprotein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5' untranslated region of this gene has been described and three variants are described, all encoding an identical protein.
TRUSTED_SUSTAINABILITY

Tekniske data

Adgangsnummer P12644
Til brug med (applikation) Control, Western Blot Control, Bioactivity
Formulering Protein with no preservative
Gen-id (Entrez) 652
Molekylvægt (g/mol) 25.6 kDa
Navn Human BMP-4
Oprensningsmetode Purified
Mængde 10 x 100 μg
Kilde HeLa cells
Opbevaringskrav -20°C
Regulatorisk status RUO
Endotoksin koncentration <1 EU/μg
Gene Alias BMP; BMP2B; BMP-2B; Bmp2b1; Bmp2b-1; Bmp4; Bmp-4; BOMPR4A; Bone morphogenetic protein; Bone morphogenetic protein 2B; bone morphogenetic protein 4; bone morphogenetic protein 4 preproprotein; DVR4; Dvr-4; MCOPS6; OFC11; ZYME
Fælles navn BMP4
Gen symbol BMP4
Biologisk aktivitet Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 2-8 ng/ml
Produkttype Protein
Konjugeret Unconjugated
Rekombinant Recombinant
Sekvens HHSQRARKKN KNCRRHSLYV DFSDVGWNDW IVAPPGYQAF YCHGDCPFPL ADHLNSTNHA IVQTLVNSVN SSIPKACCVP TELSAISMLY LDEYDKVVLK NYQEMVVEGC GCR
Indhold og opbevaring -20°C
Aktivitet Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 2-8 ng/ml.
Udtrykssystem HeLa cells
Form Lyophilized
Renhed eller kvalitetsklasse ≥ 97% by SDS-PAGE gel and HPLC analyses.
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.