missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Gibco™ Human BMP-4 Recombinant Protein, PeproTech®
Klik for at se tilgængelige muligheder
Mængde:
1 μg
10 x 100 μg
100 μg
5 μg
5 x 100 μg
50 μg
Pakningsstørrelse:
5µg
Beskrivelse
120-05-1MG will be provided as 10 x 100 μg (120-05-100UG). 120-05-500UG will be provided as 5 x 100 μg (120-05-100UG). Recombinant Human BMP-4, expressed in HeLa cells, is a 25.6 kDa homodimeric glycoprotein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5' untranslated region of this gene has been described and three variants are described, all encoding an identical protein.
Tekniske data
Tekniske data
| Adgangsnummer | P12644 |
| Til brug med (applikation) | ELISA, Functional Assay, Western Blot |
| Formulering | protein with no preservative |
| Gen-id (Entrez) | 652 |
| Molekylvægt (g/mol) | 25.6 |
| Navn | Human BMP-4 |
| Mængde | 10 x 100 μg |
| Kilde | HeLa cells |
| Opbevaringskrav | -20°C |
| Regulatorisk status | RUO |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion