Få mere at vide
R&D Systems™ Human BMP-2 DuoSet ELISA
Format:
15 96-brønds plader
5 plader med 96 brønde
Beskrivelse
The Human BMP-2 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human BMP-2.
Tekniske data
Tekniske data
| Produkttype | ELISA Kit |
| Mængde | 1 Kit |
| Arter | Human |
| Konjugeret | Biotin |
| Analyseområde | 46.90 to 3000 pg/mL |
| Krydsreaktivitet | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| Format | 5 plader med 96 brønde |
| Til brug med (applikation) | ELISA |
| Synonym | BDA2, BMP2, BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2, SSFSC |
| Indhold og opbevaring | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |