Learn More
Invitrogen™ Human beta-3 Defensin ELISA Development Kit (ABTS), PeproTech®
ELISA
Brand:  Invitrogen™ 900-K210K
Product Code. 30088274
  • 2895.00 DKK / Pack of 10
Description
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 103, which has broad spectrum antimicrobial activity and may play an important role in innate epithelial defense. [provided by RefSeq, Oct 2014].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P81534
Lot-specific
ELISA
414325
Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate
10 x 96 Tests
Refer to product documentation for component specific storage temperature.
1 hrs 45 mins
Lot-specific
HRP
Colorimetric Microplate Reader
DEFB103A
HRP
RUO
beta-3 Defensin
24 hrs 30 mins
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Invitrogen™ Human beta-3 Defensin ELISA Development Kit (ABTS), PeproTech® > 10 x 96 Tests; HRP