Learn More
Invitrogen™ Human beta-3 Defensin ELISA Development Kit (ABTS), PeproTech®
ELISA
Brand: Invitrogen™ 900-K210K
This item is not returnable.
View return policy
Description
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 103, which has broad spectrum antimicrobial activity and may play an important role in innate epithelial defense. [provided by RefSeq, Oct 2014].
Specifications
P81534 | |
Lot-specific | |
ELISA | |
414325 | |
Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate | |
10 x 96 Tests | |
Refer to product documentation for component specific storage temperature. | |
1 hrs 45 mins |
Lot-specific | |
HRP | |
Colorimetric Microplate Reader | |
DEFB103A | |
HRP | |
RUO | |
beta-3 Defensin | |
24 hrs 30 mins |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Human beta-3 Defensin ELISA Development Kit (ABTS), PeproTech® > 10 x 96 Tests; HRP
Spot an opportunity for improvement?Share a Content Correction