missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Gibco™ Human BD-2 Recombinant Protein, PeproTech®
Description
Recombinant Human BD-2 is a 4.3 kDa protein containing 41 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Defensins (alpha and beta) are cationic peptides with a broad spectrum of antimicrobial activity that comprise an important arm of the innate immune system. The alpha-defensins are distinguished from the beta-defensins by the pairing of their three disulfide bonds. To date, six human beta-defensins have been identified; BD-1, BD-2, BD-3, BD-4, BD-5 and BD-6. beta-defensins are expressed on some leukocytes and at epithelial surfaces. In addition to their direct antimicrobial activities, they can act as chemoattractants towards immature dendritic cells and memory T cells. The beta-defensin proteins are expressed as the C-terminal portion of precursors, and are released by proteolytic cleavage of a signal sequence and, in some cases, a propeptide sequence. Beta-defensins contain a six-cysteine motif that forms three intra-molecular disulfide bonds.
Specifications
Specifications
| Adgangsnummer | O15263 |
| Til brug med (applikation) | ELISA, Functional Assay, Western Blot |
| Formulering | protein with no preservative |
| Gen-id (Entrez) | 1673 |
| Molekylvægt (g/mol) | 4.3 kDa |
| Navn | Human BD-2 |
| Mængde | 100 μg |
| Kilde | E. coli |
| Regulatorisk status | RUO |
| Endotoksin koncentration | <1 EU/ μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?