Learn More
Abnova™ Human BCL2L11 Full-length ORF (NP_996885.1, 1 a.a. - 112 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010018-P02.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. The protein encoded by this gene contains a Bcl-2 homology domain 3 (BH3). It has been shown to interact with other members of the BCL-2 protein family, including BCL2, BCL2L1/BCL-X(L), and MCL1, and to act as an apoptotic activator. The expression of this gene can be induced by nerve growth factor (NGF), as well as by the forkhead transcription factor FKHR-L1, which suggests a role of this gene in neuronal and lymphocyte apoptosis. Transgenic studies of the mouse counterpart suggested that this gene functions as an essential initiator of apoptosis in thymocyte-negative selection. Several alternatively spliced transcript variants of this gene have been identified. [provided by RefSeq]
Sequence: MAKQPSDVSSECDREGRQLQPAERPPQLRPGAPTSLQTEPQDRSPAPMSCDKSTQTPSPPCQAFNHYLSAMVVILEDIGDLSLCFGFIFTGLDLYGHHHSQDTEQLNHKDFSSpecifications
NP_996885.1 | |
Liquid | |
10018 | |
BCL2L11 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BAM/BIM/BIM-alpha6/BIM-beta6/BIM-beta7/BOD/BimEL/BimL | |
BCL2L11 | |
Yes | |
wheat germ expression system |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
38.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
BCL2L11 | |
Wheat Germ (in vitro) | |
GST |