Learn More
Human BAX knockout HeLa cell lysate, 1 Kit
Product Code. 32143929 Shop All Abcam Products
Change view
Click to view available options
Unit Size:
Each
Product Code. Unit Size
32143929 Each
1 options
Product Code. 32143929 Supplier Abcam Supplier No. ab2638411Kit
 Restricted Item
Forventet forsendelse: 30-11-2026
Human BAX knockout HeLa cell/tissue lysate.
  • Species: Human
  • Tissue: BAX knockout HeLa
Specifications
For Use With (Application) Western Blot