missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Human BATF Synthetic Peptide

Artikelnummer. 15225419
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 μg
Pakningsstørrelse:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
15225419 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15225419 Leverandør Invitrogen™ Leverandørnr. PEP1505
 Begrænset vare
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Synthetic Peptide

This peptide corresponds to 16 amino acids near the amino terminus of human BATF. PEP-1505 can be used as a blocking peptide with polyclonal antibody PA5-34463.

BATF is a nuclear basic leucine zipper protein that belongs to the AP-1/ATF superfamily of transcription factors. The leucine zipper of this protein mediates dimerization with members of the Jun family of proteins. This protein is thought to be a negative regulator of AP-1/ATF transcriptional events and blocks cellular transformation by Ras and Fos. BATF also is required for the differentiation of IL-17-producing T helper (TH17) cells, a CD4+ subset of T cells that coordinates the inflammatory response in host defense. In both T cells and B cells, BATF is required for the appropriate regulation of the class-switch recombination.
TRUSTED_SUSTAINABILITY

Tekniske data

Gen-id (Entrez) 10538
Arter Human
Oprensningsmetode purified
Adgangsnummer Q16520
Fælles navn BATF
Koncentration 200 μg/mL
Indhold og opbevaring -20°C
Oversigt Synthetic Peptide
Format Liquid
Formulering PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
Til brug med (applikation) Blocking Assay, Control
Gene Alias activating transcription factor B; basic leucine zipper ATF-like transcription factor; basic leucine zipper transcription factor, ATF-like; basic leucine zipper transcription factor-like; basic leucine zipper transcriptional factor ATF-like; BATF; B-ATF; BATF1; B-cell activating transcription factor; B-cell-activating transcription factor; SFA2; SFA-2; SF-HT-activated gene 2 protein; si:ch211-153j24.4
Gen symbol Batf
Lyofiliseret, frysetørret No
Konserveringsmiddel Sodium azide
Produkttype Peptide
Mængde 50 μg
Regulatorisk status RUO
Opbevaringsbuffer PBS
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.